Learn More
Perkin Elmer US LLC Accessory Base Plate for the Fixed Angle Reflection Accessory
Supplier:  Perkin Elmer US LLC N4101030
Encompass_Preferred
Accessory Base Plates make it possible to install and use specialty accessories not normally compatible with a spectrometer. These base plates must be installed by a PerkinElmer Service Engineer.
Catalog No. 50-210-5532