Learn More
Electron Microscopy Sciences K-kits Accessory Box
SDP
Catalog No. 501928048
Encompass_Preferred
The Accessory Box contains sealing and mounting glues, four plastic sticks, and spare parts, including a spare needle, an Allen key for theNeedle Pen, a Channel Opener, and two Liquid Stages. Used with the K-kit for TEM observation of liquid samples.
Catalog No. 50-192-8048 Supplier Electron Microscopy Sciences Supplier No. K7267
The Accessory Box contains sealing and mounting glues, four plastic sticks, and spare parts, including a spare needle, an Allen key for theNeedle Pen, a Channel Opener, and two Liquid Stages. Used with the K-kit for TEM observation of liquid samples.