Learn More
Thermo Scientific™ Gateway Accessory Kit for OPC™ UA enablement of Heracell VIOS and Forma Steri-Cycle CO2 Incubators
Catalog No. 01257028
Change view
Click to view available options
For Use With (Application):
OPC UA Integration
Catalog No. For Use With (Application)
01-257-028 OPC UA Integration
1 options
Catalog No. 01-257-028 Supplier Thermo Scientific™ Supplier No. 50168119
Only null left
Add to Cart
Add to Cart
Benefit from paperless record keeping, remote monitoring, cloud analytics and more by incorporating your CO2 Incubator into your DCS (Distributed Control Systems) using the Gateway Accessory Kit for OPC™ UA enablement of the Heracell VIOS and Forma Steri-Cycle.
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Heracell VIOS and Forma Steri-Cycle CO2 Incubators using this gateway accessory kit. Benefit from the full potential of incorporating lab equipment into your new or existing DCS, including paperless record keeping, remote monitoring, cloud analytics and more.
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Heracell VIOS and Forma Steri-Cycle CO2 Incubators using this Thermo Scientific™ Gateway accessory kit. Benefit from the full potential of incorporating lab equipment into your new or existing DCS, including paperless record keeping, remote monitoring, cloud analytics and more.
Product Features
Thermo Scientific™ Gateway for OPC UA enablement of Heracell VIOS and Forma Steri-Cycle CO2 Incubators utilizes the OPC UA interoperability standard to enable a secure and reliable exchange of real-time and historical data between Distributed Control Systems (DCS) and enterprise users.
What is OPC UA?
Open Platform Communication Unified Architecture (OPC™ UA) is a platform-independent, open industry standard for connectivity that enables machine-to-machine communication and information integration between various devices, software systems, and applications.
Advantages to connecting
  • 21 CFR Part 11–compliance and GMP needs
  • Support audit requirements with enhanced data traceability
  • Real-time remote monitoring for sample protection, workflow efficiencies, asset and cost optimization, and regulatory compliance
The gateway accessory kit comes complete with a gateway device, USB 2.0 cable USB-A to USB-B connector, and 100-240V power supply. One kit is needed per CO2 Incubator connection.
TRUSTED_SUSTAINABILITY
Specifications
Description Kit available everywhere except China
CO2 Sensor Technology No Sensor
For Use With (Equipment) Heracell VIOS and Forma Steri-Cycle CO2 Incubators
Lighting No Lighting
Material Polystyrene, Polyethylene
Oxygen Control None
Plug Type Power supply with mains plug 100-240V
Type Gateway Device for OPC UA
For Use With (Application) OPC UA Integration
Electrical Requirements Power supply with mains plug 100-240V
Voltage 100/240 V