Learn More
TA Instruments Accessory Kit Discovery DSC 25/250/2500, Includes Tweezers, Indium Calibration, Tzero Sample Pans and Lids, Hermetic Lids, Sapphire Calibration Kit, DSC Cleaning Brush
Catalog No. 5000580648 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Accessory Kit Discovery DSC 25/250/2500 includes stainless steel tweezers, indium calibration material, Tzero sample pans and lids, Tzero hermetic lids, MDSC sapphire calibration kit, DSC cleaning brush, and Tzero sapphire calibration kit.
Catalog No. 50-005-80648 Supplier TA Instruments Supplier No. 973011.901
Waters TA Instruments Accessory Kit Discovery DSC 25/250/2500 includes stainless steel tweezers, indium calibration material, Tzero sample pans and lids, Tzero hermetic lids, MDSC sapphire calibration kit, DSC cleaning brush, and Tzero sapphire calibration kit.