Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ Accessory for PIPETMAN™ Complete Pipetting System Carrousel™ Pipet Stand PROMO

Catalog No. F161401GPM Shop All Gilson Products
Change view
Click to view available options
Gilson™ Complete Pipetting System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
F161401GPM Gilson™ Complete Pipetting System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F161401GPM Supplier Gilson™ Supplier No. F161401PM

Convenient rotating stand holds up to seven single and multichannel Gilson pipets.

  • Robust, stable and space saving stand
  • Makes bench work look neat and tidy
  • Heavy base of stand prevents toppling over

Specifications

Color White
Type Pipette Stand
For Use With (Equipment) PIPETMAN Complete Pipetting System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.