Learn More
New England Biolabs, Inc. AccI – 1000 units
SDP
Catalog No. 50812064
Encompass
A restriction endonuclease that recognizes the sequence GT^MK_AC.
Catalog No. 50-812-064 Supplier New England Biolabs, Inc. Supplier No. R0161S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GT^MK_AC.