Learn More
New England Biolabs, Inc. AccI – 5000 units
SDP
Catalog No. 50812063
Encompass
A restriction endonuclease that recognizes the sequence GT^MK_AC.
Catalog No. 50-812-063 Supplier New England Biolabs, Inc. Supplier No. R0161L
A restriction endonuclease that recognizes the sequence GT^MK_AC.