Learn More
Cole-Parmer Oakton ATC Probe for 150-, 300-, 450-series meters
Supplier:  Cole-Parmer 3561805
Encompass_Preferred
Oakton ATC Probe for 150-, 300-, 450-series meters, Provides ATC for most current Oakton pH meters, Corrosion-resistant stainless steel 30 K thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100C.
Catalog No. NC9581204