Learn More
Aviva Systems Biology Corp ACCN1 Antibody - C-terminal region (ARP35033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ACCN1. Alternate names: ACCN, BNC1, MDEG, ACCN1, BNaC1, ASIC2a, hBNaC1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55633