Learn More
Aviva Systems Biology Corp ACCN1 Antibody - middle region (ARP35537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACCN1. Alternate names: ACCN, ASIC2, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1, ACCN1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55776