Learn More
Aviva Systems Biology Corp ACCN2 Antibody - N-terminal region (ARP35298_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35298_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACCN2. Alternate names: ASIC, ASIC1A, BNaC2, hBNaC2, ACCN2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55691