Learn More
Aviva Systems Biology Corp Accn2 Antibody - N-terminal region (ARP35035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Accn2. It was validated on Western Blot. . Alternate names: AI843610, ASIC, ASIC1, ASIC1a, B530003N02Rik, BNaC2, Accn2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-439