Learn More
Aviva Systems Biology Corp ACCN3 Antibody - N-terminal region (ARP35152_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35152_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ACCN3. Alternate names: ACCN3, TNaC1, DRASIC, SLNAC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55653