Learn More
Aviva Systems Biology Corp ACCN4 Antibody - middle region (ARP35288_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35288_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACCN4. Alternate names: ASIC4, BNAC4, MGC17248, MGC24860, ACCN4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63077