Learn More
Aviva Systems Biology Corp ACCN5 Antibody - middle region (ARP37696_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37696_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACCN5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACCN5. Alternate names: INAC, HINAC, ACCN5ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-56399