Learn More
Felix Storch Inc ACCUCOLD Accucold Compact Refrigerator 2.5 Cubic Feet
SDP
Catalog No. NC2158074
Encompass_Preferred
The 2.5 cu.ft. FF31L7 utilizes automatic defrost operation for low maintenance use. Four removable chrome shelves offer convenient storage, while the flat door liner allows you to store deeper items. The digital thermostat is located inside and can be managed in Celsius or Fahrenheit. Interior LED lighting offers an attractive, energy efficient view of the interior in a soft blue hue: https://www.accucold.com/product/FF31L7
This item is not returnable. View return policy
Catalog No. NC2158074 Supplier Felix Storch Inc Supplier No. FF31L7
The 2.5 cu.ft. FF31L7 utilizes automatic defrost operation for low maintenance use. Four removable chrome shelves offer convenient storage, while the flat door liner allows you to store deeper items. The digital thermostat is located inside and can be managed in Celsius or Fahrenheit. Interior LED lighting offers an attractive, energy efficient view of the interior in a soft blue hue: https://www.accucold.com/product/FF31L7