Learn More
Electron Microscopy Sciences Flex-Tainer™ Accordion-style Jug 5 L
SDP
Supplier:  Electron Microscopy Sciences 6422815
Encompass_Preferred
Made of high strength polypropylene this space-saving expandable accordion-style jug flattens down for easy transport and storage. Great ideal for storing and carrying numerous types of fluids, it is suitable for any laboratory. Features a comfortable carry grip and screw-on cap with on/off spout. Comes individually wrapped.
Catalog No. 50-190-0263