Learn More
Waters Corp Waters Reagent Kit, Amino Acid, LC, Reversed Phase, Optical
Catalog No. 50785689 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The AccQ-Fluor Reagent Kit offers a highly efficient and reliable method for amino acid analysis. Designed for HPLC/UHPLC systems, it provides a streamlined and sensitive approach for derivatization and analysis. The kit utilizes a unique derivatization agent, reacting rapidly and quantitatively to produce stable, highly fluorescent derivatives for consistent, reproducible results. Its versatility supports biological, food, and beverage sample analysis.
  • Highly efficient and reliable amino acid detection.
  • Designed for use with HPLC and UHPLC systems.
  • Streamlined and sensitive derivatization and analysis.
  • Exceptional sensitivity for trace amino acid quantification.
  • Minimizes background interference from sample matrix.
  • Versatile for biological, food, and beverage sample analysis.
Catalog No. 50-785-689 Supplier Waters Corp Supplier No. WAT052880
The AccQ-Fluor Reagent Kit offers a highly efficient and reliable method for amino acid analysis. Designed for HPLC/UHPLC systems, it provides a streamlined and sensitive approach for derivatization and analysis. The kit utilizes a unique derivatization agent, reacting rapidly and quantitatively to produce stable, highly fluorescent derivatives for consistent, reproducible results. Its versatility supports biological, food, and beverage sample analysis.
  • Highly efficient and reliable amino acid detection.
  • Designed for use with HPLC and UHPLC systems.
  • Streamlined and sensitive derivatization and analysis.
  • Exceptional sensitivity for trace amino acid quantification.
  • Minimizes background interference from sample matrix.
  • Versatile for biological, food, and beverage sample analysis.