Learn More
Waters Corp Waters AccQ-Tag Chemistry Kit, 250 Analyses, HPLC, UV, Fluorescence
Supplier:  Waters Corp WAT052875
Encompass_Preferred
The kit is used for Amino Acid Analysis (AAA) and is designed for up to 250 analyses. It supports HPLC and UHPLC systems with UV and fluorescence detection. This chemistry package provides reagents and documentation for the analysis of protein and peptide hydrolysate amino acids.
  • Amino acid analysis is required for many applications.
  • Includes various reagents, a column, sample tubes, an amino acid standard, and a user guide.
  • Designed for 250 analyses.
  • Supports HPLC and UHPLC separation modes.
  • Detection methods include UV and fluorescence.
  • Storage condition is room temperature.
  • Reagent (6-aminoquinolyl-N-hydroxysuccinimidyl carbamate or AQC) derivatizes primary and secondary amines in a single-step reaction.
  • Produces highly stable, fluorescent adducts.
  • High purity reagents minimize background amino acid content.
Catalog No. 50-785-688