Learn More
Waters Corp Waters AccQ-Tag Eluent A, Reversed Phase, Optical Detection, 1 Liter
Supplier:  Waters Corp WAT052890
Encompass_Preferred
AccQ-Tag Eluent A is a proprietary buffer solution optimized for high-performance liquid chromatography (HPLC) amino acid analysis. Its precise composition ensures consistent pH and ionic strength, critical for reliable retention times and optimal resolution. This product is used for accurate identification and quantification of amino acids in biological research, food, beverage, feed, and biotherapeutic development.
  • Optimized for amino acid analysis (AAA) using HPLC systems with fluorescence detection.
  • Overcomes separation and detection difficulties for a wide range of chemical properties.
  • Provides accurate, precise, and consistent quantitative results.
  • Offers rugged derivatization chemistry, stable chromatographic baselines, and superior amino acid resolution.
  • Ensures robust routine analyses across various applications.
Catalog No. 50-785-691