Learn More
Waters Corp Waters AccQ-Tag Ultra Chemistry Kit, 1.7 μm C18, ACQUITY Separations
Supplier:  Waters Corp 176001235
Encompass_Preferred
This chemistry kit is designed for the separation of hydrolysate amino acid derivatives on ACQUITY LC Systems. It includes a column, pre-packaged reagents, and QC-tested eluents for pre-column derivatization of primary and secondary amines, yielding stable adducts quantifiable by UV detection.
  • Includes a column, pre-packaged reagents, and QC-tested eluents.
  • Column contains ethylene-bridged hybrid (BEH) C18, 130 Å, 1.7 μm particles.
  • Batch-tested for consistent performance in amino acid analysis.
  • Features eCord intelligent chip technology for lifetime performance tracking and data capture.
Catalog No. 50-780-045