Learn More
Waters Corp Waters AccQ·Tag Ultra RP Column, C18, 130Å, 1.7 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186003837
Encompass_Preferred
The AccQ·Tag Ultra RP Column is designed for reversed-phase separation of amino acids derivatized with AccQ·Tag Ultra reagent. Each column is quality control tested using an amino acid standard for repeatable performance in demanding amino acid analysis. This 1.7 μm, 2.1 mm x 100 mm column is an integral part of a UPLC Amino Acid Analysis Solution.
  • Specifically designed for reversed-phase separation of derivatized amino acids.
  • Quality control tested using an amino acid standard for repeatable performance.
  • Accomplishes accurate, high-sensitivity UPLC analyses.
  • Utilizes UPLC technology with 1.7 μm particles for improved sensitivity, resolution, and speed.
  • Enables derivatization of both primary and secondary amino acids using a single step.
  • Optimized for use with the AccQ·Tag Ultra Chemistry package.
Catalog No. 50-782-040