Learn More
Waters Corp Waters Corporation AccQ-Tag Ultra Derivatization Kit, UV Detection, 250 Analyses
Supplier:  Waters Corp 186003836
Encompass_Preferred
This derivatization kit is a component of an amino acid analysis application solution, specifically designed for amino acid analysis with UV detection. It includes five sets of reagents for 250 analyses, providing accurate results across various applications.
  • Designed for amino acid analysis with UV detection.
  • Provides reagents for 250 analyses.
  • Includes borate buffer, 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate reagent, and acetonitrile diluent.
  • Allows derivatization without special sample preservation and tolerates buffer salts.
  • Samples remain stable for several days post-preparation.
  • Delivers accurate results for protein hydrolysates and cell culture media monitoring.
  • Utilizes HPLC for fast derivatization (under 10 minutes), enhancing sensitivity and stability.
  • Compatible with biological samples: plasma, urine, tissue, and cell cultures.
Catalog No. 50-782-039