Learn More
Waters Corp Waters Corporation Eluent, 1 L, Mobile Phase, Amino Acid
Supplier: Waters Corp 186003838
This AccQ·Tag Ultra Eluent is a 1 L mobile phase for accurate, high-sensitivity UPLC amino acid analyses, enabling single-step derivatization of primary and secondary amino acids with quantitative results.
- Accurate, high-sensitivity UPLC amino acid analyses.
- Single-step derivatization of amino acids.
- Quantitative results.
- Part of a complete UPLC amino acid analysis solution.
- Suitable for protein/peptide hydrolysates, cell culture media, and nutritional analysis.
- Controls variability.
- Rugged, reliable method.
- Storage: Room temperature.
- Volume: 1 L.
- Type: Mobile phase.
- Application: Amino acid.
- Units: 1 per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.