Learn More
Waters Corp Waters AccQ-Tag Ultra Eluent B, 1 L, Amino Acid Analysis
Supplier:  Waters Corp 186003839
Encompass_Preferred
AccQ-Tag Ultra Chemistry provides a leading chemical solution for amino acid analysis, specifically designed for precise and high-sensitivity UPLC analysis. This eluent is part of a complete chemical package of fully evaluated reagents, columns, and eluents, enabling quick and easy derivatization of primary and secondary amino acids in a single step, yielding quantitative results.
  • Versatile and proven method for amino acid analysis.
  • Designed for precise and high-sensitivity UPLC analysis.
  • Uses AccQ-Fluor™ reagent for quick and easy derivatization of primary and secondary amino acids in a single step.
  • Generates unparalleled quantitative results.
  • Complete set of fully evaluated reagents, columns, and eluents.
  • Suitable for a greater number of free amino acids found in process samples, such as cell cultures or fermentation broths.
Catalog No. 50-782-042