Learn More
CAS DATALOGGERS Accsense Wi-Fi Pro Temperature Data Logger with Phone Call/SMS/Email/App Alarming, 1 Yr Cloud Subscription, and Accessories
Catalog No. NC3847154
Encompass
"Refrigerator and Freezer Datalogger Kit with 1 Year of Accsense Pro Monitoring (includes data storage, audit trail reports, multiple users, and alarms via SMS, Phone Call/Voice, Email, and App Notifications).Ethernet Temperature Data Logger with 3 Analog and 1 Digital Inputs, RTD Probe (-100C to 100C), E1-34 Nylon Thermal Buffer, and Power Supply. Calibrated at 1 Standard Point between -20C to 123C"
Catalog No. NC3847154 Supplier CAS DATALOGGERS Supplier No. ACCKITWIFI1CAL
"Refrigerator and Freezer Datalogger Kit with 1 Year of Accsense Pro Monitoring (includes data storage, audit trail reports, multiple users, and alarms via SMS, Phone Call/Voice, Email, and App Notifications).Ethernet Temperature Data Logger with 3 Analog and 1 Digital Inputs, RTD Probe (-100C to 100C), E1-34 Nylon Thermal Buffer, and Power Supply. Calibrated at 1 Standard Point between -20C to 123C"