Learn More
Hamilton Thorne Inc ACCU-BEADS
SDP
Catalog No. NC0136046
Encompass_Preferred
5 ml, latex bead suspension in aqueous medium. Contains extremely dilute sodium azide preservative [0.07%] and small amounts of propane-triol and propane I-2diol. Store at room temperature with bottle tightly capped.
Catalog No. NC0136046 Supplier Hamilton Thorne Inc Supplier No. HT710110
5 ml, latex bead suspension in aqueous medium. Contains extremely dilute sodium azide preservative [0.07%] and small amounts of propane-triol and propane I-2diol. Store at room temperature with bottle tightly capped.