Learn More
ACCU-SCOPE™ ACCU-CAM 500 EP Eyepiece Camera
SDP
Catalog No. 01189218 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
USB Microscope Camera
Catalog No. Type
01-189-218 USB Microscope Camera
1 options
Catalog No. 01-189-218 Supplier ACCU-SCOPE™ Supplier No. AU500EP
Add to Cart
Add to Cart
The affordable ACCU-CAM 500 produces 5 MP color images and video. USB 2.0 connectivity and advanced ACCU-VIEW imaging software for camera controls, live viewing, image capture and measurement. Features 5.1 MP CMOS color eyepiece camera and high speed USB 2.0 mini cable.
  • Perfect solution for those on a tight budget who need to capture images and video!
  • 1/2.8 in. SONY IMX335 color camera sensor
  • 5.1MP CMOS eyepiece camera
  • USB 2.0 direct connect to your PC
  • Includes our feature-rich ACCU-View software (PC only) for image analysis, annotation, measurement, white balance and more! Or try our ACCU-View LITE (for PC and Mac) for a streamlined operating experience (e.g., fewer features, simple interface).
  • Captures digital still and video
  • Fits into a 23.2 mm I.D. eye tube or photo tube. Optional adapter (CAT #5036) available for 30 mm I.D. eyepieces
  • In the box: ACCU-CAM 500 eyepiece camera, USB 2.0 mini cable, ACCU-View software USB stick
Specifications
Type USB Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile