Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ ACCU-CAM WiFi Camera
GREENER_CHOICESDP

Catalog No. 01189210 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
5 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-189-210 5 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-210 Supplier ACCU-SCOPE™ Supplier No. AUACWIFI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ACCU-CAM WiFi Microscope Camera delivers excellent color images and video directly to your mobile device via WiFi signals and the AccuView mobile app - no need for a computer.

  • Compact and convenient full HD imaging using just your smartphone!
  • Runs on Windows PC, Android, and Apple (iOS)
  • WiFi or USB connectivity
  • ACCU-CAM WiFi cameras are compatible with AccuView apps for PC, Android, and Apple devices. Download the mobile apps on Google Play and Apple App Store.
  • 2 MP SONY Color CMOS sensor
  • Built-in 2.4GHz WiFi adapter, IEEE802.11ac protocol
  • Capture images and video, and view them with the AccuView mobile or PC app. For a streamlined experience, try AccuView LITE for Windows and MacOS
  • View live images at up to 50 frames per second (1920 x 1080)
  • 1/2.3 in. camera sensor format - 0.35x c-mount adapter recommended
  • Connect up to 3 mobile devices simultaneously to a single ACCU-CAM WiFi camera! (up to 3 cameras can operate simultaneously in a classroom environment or over local area network)
  • Advanced camera control, image analysis, annotation, measurement, etc. with AccuView Windows PC software (free: available to download)
  • Please review the brochure for Windows PC, Macintosh, Apple (iOS) and Android system requirements

Specifications

Type Digital WiFi Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 5 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.