Learn More
BRANDTECH™ BRAND™ Accessories for accu-jet™ Pipette Controller
GSA_VA
Catalog No. 1387952 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Type:
NiMH Battery
Silicone Adapter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-879-52 Silicone Adapter
13-688-567 NiMH Battery
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-879-52 Supplier BRANDTECH™ BRAND™ Supplier No. 26508
Add to Cart
Add to Cart
Accessories and replacement parts for use with your BRAND™ accu-jet™ S and pro pipette controller.
For accu-jet™ S:
  • Part No. 26539: Plastic wall support
  • Part No. 26606: Universal AC Adapter, 100-240V / 50-60Hz
  • Part No. 26547: Snape tape
For accu-jet™ pro:
  • Part No. 26540: Plastic wall support
For accu-jet™ S and accu-jet™ S:
  • Part No: 26630: NiMH Battery
  • Part No. 26508: Silicone Adapter, with non-return valve, 54 mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With accu-jet S
Material Silicone
Type Silicone Adapter
For Use With (Equipment) accu-jet S and pro Pipet Controller
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov