Learn More
LAMDA BIOTECH INC Accu Prestained Prot Ladder II
Catalog No. NC3724890
Encompass_Preferred
The Accu™ Prestained Protein Ladder II is a ready-to-use protein molecular weight marker containing 10 prestained proteins spanning 8.5kDa to 180kDa. Engineered for protein visualization during electrophoresis and Western blot procedures, this ladder features distinctive red (70kDa) and green (25kDa) reference bands for easy molecular weight determination.
Catalog No. NC3724890 Supplier LAMDA BIOTECH INC Supplier No. G02
The Accu™ Prestained Protein Ladder II is a ready-to-use protein molecular weight marker containing 10 prestained proteins spanning 8.5kDa to 180kDa. Engineered for protein visualization during electrophoresis and Western blot procedures, this ladder features distinctive red (70kDa) and green (25kDa) reference bands for easy molecular weight determination.