Learn More
Electron Microscopy Sciences Accu-Punch Disposable Skin Biopsy Punch 5.0 mm
SDP
Supplier:  Electron Microscopy Sciences 690380550
Encompass_Preferred
The only disposable punch made in the USA. Sterile, disposable skin biopsy punches. Available in 14 different sizes as well as assortment packs. Always sharp, always sterile, ergonomic design, and ribbed for maximum control. Available in Dispenser box.
Catalog No. 50-366-769