Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemosure™ Accu-Reader™ A100 Control Set
SDP

Catalog No. 23800590
Change view
Click to view available options
Type:
iFOB External Control Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-800-590 iFOB External Control Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-590 Supplier Hemosure™ Supplier No. AR01TC01

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Control set that comes with 2 mL negative control materials and 2 mL positive control materials for use with Accu-Reader™ A100 Analyze.

  • Control Set includes: 2 mL negative control material and 2 mL positive control material
  • Storage condition: 2°C to 8°C

Specifications

CE Marker Yes
Clia Complexity Moderate
Format Tube
Sample Type Fecal Occult Blood
Type iFOB External Control Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.