Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemosure™ Accu-Reader™ A100 Sample Collection Tube
SDP

Catalog No. 23800592
Change view
Click to view available options
Type:
Sample Collection Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-800-592 Sample Collection Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-592 Supplier Hemosure™ Supplier No. AR01TB50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sample Collection Tube for use with Accu-Reader™ A100 Analyzer.

  • Fecal sample collection tube with 2 mL iFOB buffer solution
  • Storage condition: 4°C to 30°C

Specifications

CE Marker Yes
Clia Complexity Not Applicable
Format Tube
Type Sample Collection Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.