Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemosure™ Accu-Reader™ A100 Test Cartridge (iFOB)
SDP

Catalog No. 23800591
Change view
Click to view available options
Type:
iFOB Test Cartridges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-800-591 iFOB Test Cartridges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-591 Supplier Hemosure™ Supplier No. AR01CT50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Individually sealed iFOB immunoassay test cartridge for use with Accu-Reader™ A100 Analyzer.

  • Individually sealed iFOB immunoassay test cartridge
  • Storage condition: 4°C to 30°C

Specifications

CE Marker Yes
Clia Complexity Moderate
Sensitivity C50=100 ng/mL
Type iFOB Test Cartridges
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.