Learn More
Hamilton Thorne Inc Accu-Beads Triple Package, Quality Control Beads, 3 concentrations (3 M/ml, 18 M/ml, 35 M/ml), 5 ml per aliquot, Static (non-motile), Room temperature storage, Shelf Life 2 years
SDP
Catalog No. NC1451157
Encompass_Preferred
Accu-Beads Triple Package, Quality Control Beads, 3 concentrations (3 M/ml, 18 M/ml, 35 M/ml), 5 ml per aliquot, Static (non-motile), Room temperature storage, Shelf Life 2 years
Catalog No. NC1451157 Supplier Hamilton Thorne Inc Supplier No. 710111
Accu-Beads Triple Package, Quality Control Beads, 3 concentrations (3 M/ml, 18 M/ml, 35 M/ml), 5 ml per aliquot, Static (non-motile), Room temperature storage, Shelf Life 2 years