Learn More
Labnet International, Inc Labnet Digital Dry Bath, Single Block, 120V
Catalog No. NC1296883
Encompass_Preferred
The Labnet Digital Dry bath incubators are available in single, dual, and four block configurations and provide a broad temperature range up to 150°C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories. They have been designed to incorporate the best combination of features including block capacity, dual displays, and an external temperature probe that gives life science researchers the confidence and efficiency they need when working with precious samples.
Catalog No. NC1296883 Supplier Labnet International, Inc Supplier No. D1301
The Labnet Digital Dry bath incubators are available in single, dual, and four block configurations and provide a broad temperature range up to 150°C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories. They have been designed to incorporate the best combination of features including block capacity, dual displays, and an external temperature probe that gives life science researchers the confidence and efficiency they need when working with precious samples.