Learn More
Biotium AccuBlue Broad Range dsDNA Quantitation Solution, trial size
Supplier:  Biotium 31009T
Encompass_Preferred
The AccuBlue Broad Range dsDNA Quantitation Solution offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. The solution provides a linear detection range from 2 – 2000 ng and is compatible with fluorescence microplate readers. The solution has an excitation/emission of 350/460 nm. This item is a standalone fluorescent DNA quantitation solution, and does not include DNA standards. It is intended for users who wish to use their own DNA standards. This trial size has sufficient reagent for 200 assays.
Catalog No. 50-196-4534