Learn More
Biotium AccuBlue Broad Range dsDNA Quantitation Kit with DNA Standard, trial size (200 assays)
Catalog No. 501964531
Encompass_Preferred
The AccuBlue Broad Range dsDNA Quantitation Kit offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. The kit provides a linear detection range from 2 – 2000 ng and is compatible with fluorescence microplate readers. The solution has an excitation/emission of 350/460 nm. This trial size kit has sufficient reagent for 200 assays.
Catalog No. 50-196-4531 Supplier Biotium Supplier No. 31007T
The AccuBlue Broad Range dsDNA Quantitation Kit offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. The kit provides a linear detection range from 2 – 2000 ng and is compatible with fluorescence microplate readers. The solution has an excitation/emission of 350/460 nm. This trial size kit has sufficient reagent for 200 assays.