Learn More
Biotium AccuBlue Broad Range RNA Quantitation Kit, Trial size (200 assays)
Catalog No. 501964584
Encompass_Preferred
The AccuBlue Broad Range RNA Quantitation Kit boasts the widest linear range (5¿1000 ng RNA) of available RNA quantitation kits, while providing exceptional accuracy, sensitivity, and high RNA selectivity. A mammalian RNA standard is included for accurate quantitation of mammalian cell RNA. The assay is linear between 5 and 1000 ng of RNA per 200 uL assay well, which spans both Qubit HS and Qubit BR ranges. Unlike absorbance-based measurements, RNA Broad Range Dye is selective for RNA over double-stranded DNA, and can tolerate an equimolar amount of dsDNA in the sample without significant effect on RNA quantitation. The AccuBlue Broad Range RNA Quantitation assay has an excitation/emission of 650/670 nm (when bound to RNA) and is suitable for fluorescence microplate readers and for the Qubit fluorometer. This trial size kit has sufficient reagent for 200 assays.
Catalog No. 50-196-4584 Supplier Biotium Supplier No. 31073T
The AccuBlue Broad Range RNA Quantitation Kit boasts the widest linear range (5¿1000 ng RNA) of available RNA quantitation kits, while providing exceptional accuracy, sensitivity, and high RNA selectivity. A mammalian RNA standard is included for accurate quantitation of mammalian cell RNA. The assay is linear between 5 and 1000 ng of RNA per 200 uL assay well, which spans both Qubit HS and Qubit BR ranges. Unlike absorbance-based measurements, RNA Broad Range Dye is selective for RNA over double-stranded DNA, and can tolerate an equimolar amount of dsDNA in the sample without significant effect on RNA quantitation. The AccuBlue Broad Range RNA Quantitation assay has an excitation/emission of 650/670 nm (when bound to RNA) and is suitable for fluorescence microplate readers and for the Qubit fluorometer. This trial size kit has sufficient reagent for 200 assays.