Learn More
Biotium AccuBlue High Sensitivity dsDNA Standards, set of eight, 0.5 mL each
Catalog No. 501964529
Encompass_Preferred
Stand-alone set of the eight double-stranded DNA standards provided with the AccuBlue High Sensitivity dsDNA Quantitation Kit. Calf thymus dsDNA in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide. Set of eight 0.5 mL standards at 0, 0.5, 1, 2, 4, 6, 8 and 10 ng/uL.
Catalog No. 50-196-4529 Supplier Biotium Supplier No. 31006C
Stand-alone set of the eight double-stranded DNA standards provided with the AccuBlue High Sensitivity dsDNA Quantitation Kit. Calf thymus dsDNA in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide. Set of eight 0.5 mL standards at 0, 0.5, 1, 2, 4, 6, 8 and 10 ng/uL.