Learn More
Biotium AccuBlue High Sensitivity dsDNA Quantitation Kit
Catalog No. 501964528
Encompass_Preferred
AccuBlue High Sensitivity dsDNA Quantitation Assay is a safer fluorescent dsDNA quantitation assay for microplate reader. The assay uses non-toxic, non-mutagenic EvaGreen dye. Featuring our unique AccuBlue High Sensitivity Enhancer, the assay provides highly sensitive and selective detection of purified dsDNA samples with excellent linearity. This kit includes calf thymus dsDNA standards, and provide sufficient reagents to perform 1000 assays based on a 96-well microplate format. The assay is compatible with microplate readers, spectrofluorometers and hand-held minifluorometers. Note that while the assay is spectrally compatible with Qubit fluorometers from Thermo Fisher, it may not be compatible with the high sensitivity dsDNA program on all Qubit models. Best linearity is achieved at an excitation/emission of 485/530 nm.
Catalog No. 50-196-4528 Supplier Biotium Supplier No. 31006
AccuBlue High Sensitivity dsDNA Quantitation Assay is a safer fluorescent dsDNA quantitation assay for microplate reader. The assay uses non-toxic, non-mutagenic EvaGreen dye. Featuring our unique AccuBlue High Sensitivity Enhancer, the assay provides highly sensitive and selective detection of purified dsDNA samples with excellent linearity. This kit includes calf thymus dsDNA standards, and provide sufficient reagents to perform 1000 assays based on a 96-well microplate format. The assay is compatible with microplate readers, spectrofluorometers and hand-held minifluorometers. Note that while the assay is spectrally compatible with Qubit fluorometers from Thermo Fisher, it may not be compatible with the high sensitivity dsDNA program on all Qubit models. Best linearity is achieved at an excitation/emission of 485/530 nm.