Learn More
Biotium AccuBlue High Sensitivity dsDNA Quantitation Solution
Catalog No. 501964532
Encompass_Preferred
The AccuBlue High Sensitivity dsDNA Quantitation Solution offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. The solution provides sufficient reagents to perform 1000 assays based on a 96-well microplate format. This item is a standalone fluorescent DNA quantitation solution, and is intended for customers who wish to use their own DNA standards. We also sell the complete AccuBlue High Sensitivity Quantitation Kit that includes a set of standards. Best linearity is achieved at an excitation/emission of 485/530 nm.
Catalog No. 50-196-4532 Supplier Biotium Supplier No. 31008
The AccuBlue High Sensitivity dsDNA Quantitation Solution offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. The solution provides sufficient reagents to perform 1000 assays based on a 96-well microplate format. This item is a standalone fluorescent DNA quantitation solution, and is intended for customers who wish to use their own DNA standards. We also sell the complete AccuBlue High Sensitivity Quantitation Kit that includes a set of standards. Best linearity is achieved at an excitation/emission of 485/530 nm.