Learn More
Biotium AccuBlue High Sensitivity dsDNA Quantitation Solution, trial size
Supplier:  Biotium 31008T
Encompass_Preferred
The AccuBlue High Sensitivity dsDNA Quantitation Solution offers sensitive and selective detection of purified dsDNA samples with minimal effects from common contaminants. This trial size provides sufficient reagents to perform 200 assays based on a 96-well microplate format. This item is a standalone fluorescent DNA quantitation solution, and is intended for customers who wish to use their own DNA standards. We also sell the complete AccuBlue High Sensitivity Quantitation Kit that includes a set of standards. Best linearity is achieved at an excitation/emission of 485/530 nm.
Catalog No. 50-196-4533