Learn More
Biotium AccuBlue NextGen dsDNA Quantitation Solution, Trial Size (200 assays)
Catalog No. 501964571
Encompass_Preferred
The AccuBlue NextGen dsDNA Quantitation Solution provides an unprecedented level of sensitivity in dsDNA detection. The AccuBlue NextGen dsDNA Quantitation assay is the most sensitive and accurate DNA quantitation method for ultra low amounts of DNA. With a detection limit as low as 1 pg DNA in a 96-well plate reader assay, AccuBlue NextGen outperforms rivals such as PicoGreen in sensitivity and accuracy. Specially designed for scientists who need to quantify extremely dilute or precious DNA, AccuBlue NextGen is ideal for use in preparing samples for applications such as Next Generation Sequencing (NGS) and digital PCR. The AccuBlue NextGen dsDNA Quantitation Solution does not come with any DNA standards. It is intended customers who wish to use their own standards. The linear detection range of the assay is 1-3000 pg dsDNA. This trial size has sufficient reagent for 200 assays.
Catalog No. 50-196-4571 Supplier Biotium Supplier No. 31061T
The AccuBlue NextGen dsDNA Quantitation Solution provides an unprecedented level of sensitivity in dsDNA detection. The AccuBlue NextGen dsDNA Quantitation assay is the most sensitive and accurate DNA quantitation method for ultra low amounts of DNA. With a detection limit as low as 1 pg DNA in a 96-well plate reader assay, AccuBlue NextGen outperforms rivals such as PicoGreen in sensitivity and accuracy. Specially designed for scientists who need to quantify extremely dilute or precious DNA, AccuBlue NextGen is ideal for use in preparing samples for applications such as Next Generation Sequencing (NGS) and digital PCR. The AccuBlue NextGen dsDNA Quantitation Solution does not come with any DNA standards. It is intended customers who wish to use their own standards. The linear detection range of the assay is 1-3000 pg dsDNA. This trial size has sufficient reagent for 200 assays.