Learn More
Alkali Scientific AccuCheck Pipette Calibration
SDP
Supplier:  Alkali Scientific RS7537
Encompass_Preferred
The new AccuCheck Pipette Calibration System (W3002A-120 Balance with W3110 Anti Evaporation Trap) gives your lab the freedom to calibrate pipettes quickly and when required The system combines our popular Tx touch screen analytical balance with an anti evaporation trap accessory and built-in pipette calibration software Together this standalone pipette calibration system is easy for any lab tech to use for the ISO 8655 method of pipette calibration The user interface of the AccuChecksoftware is intuitive and requires no special training to use Individual pipette details (brand model serial number) can be saved in the internal data base for current and future calibrations The software walks the user through all steps of the gravimetric method allows input of water temp air temp pressure and humidity It saves the weight values and then performs all calculations for accuracy and precision at 3 volume settings The Anti Evaporation Trap greatly improves results when performing t
Catalog No. 50-302-9581