Learn More
Biotium AccuClear Ultra High Sensitivity dsDNA Quantitation Kit with DNA Standard, trial size (200 assays)
Catalog No. 501964546
Encompass_Preferred
AccuClear Ultra High Sensitivity dsDNA Quantitation Kit utilizes a novel, next-generation DNA binding dye for highly sensitive and accurate quantitation of purified dsDNA samples across a broad range of DNA concentrations. The assay is linear between 0.03 ng and 250 ng of dsDNA per assay in microplate format, making it the most versatile DNA quantitation assay. Unlike absorbance-based measurements, AccuClear dye is highly selective for double-stranded DNA over single stranded DNA or RNA. The AccuClear dsDNA quantitation assay is designed for use with fluorescence 96-well plate readers equipped with excitation and emission filters for detecting green fluorescence. The unique spectral properties of AccuClear dye make it especially well-suited for use with instruments with blue LED excitation sources. This trial size kit has sufficient reagent for 200 assays.
Catalog No. 50-196-4546 Supplier Biotium Supplier No. 31028T
AccuClear Ultra High Sensitivity dsDNA Quantitation Kit utilizes a novel, next-generation DNA binding dye for highly sensitive and accurate quantitation of purified dsDNA samples across a broad range of DNA concentrations. The assay is linear between 0.03 ng and 250 ng of dsDNA per assay in microplate format, making it the most versatile DNA quantitation assay. Unlike absorbance-based measurements, AccuClear dye is highly selective for double-stranded DNA over single stranded DNA or RNA. The AccuClear dsDNA quantitation assay is designed for use with fluorescence 96-well plate readers equipped with excitation and emission filters for detecting green fluorescence. The unique spectral properties of AccuClear dye make it especially well-suited for use with instruments with blue LED excitation sources. This trial size kit has sufficient reagent for 200 assays.