Learn More
Biotium AccuClear Ultra High Sensitivity dsDNA Quantitation Solution (1000 assays)
Catalog No. 501964543
Encompass_Preferred
AccuClear Ultra High Sensitivity dsDNA Quantitation Solution utilizes a novel, next-generation DNA binding dye for highly sensitive and accurate quantitation of purified dsDNA samples across a broad range of DNA concentrations. The assay is linear between 0.03 ng and 250 ng of dsDNA per assay in microplate format making it the most versatile DNA quantitation assay. Unlike absorbance-based measurements, AccuClear dye is highly selective for double-stranded DNA over single stranded DNA or RNA. This product is a standalone AccuClear Quantitation Solution that does not come with any DNA standards, intended for users that wish to use their own DNA standards. This product has sufficient reagent for 1000 assays.
Catalog No. 50-196-4543 Supplier Biotium Supplier No. 31027
AccuClear Ultra High Sensitivity dsDNA Quantitation Solution utilizes a novel, next-generation DNA binding dye for highly sensitive and accurate quantitation of purified dsDNA samples across a broad range of DNA concentrations. The assay is linear between 0.03 ng and 250 ng of dsDNA per assay in microplate format making it the most versatile DNA quantitation assay. Unlike absorbance-based measurements, AccuClear dye is highly selective for double-stranded DNA over single stranded DNA or RNA. This product is a standalone AccuClear Quantitation Solution that does not come with any DNA standards, intended for users that wish to use their own DNA standards. This product has sufficient reagent for 1000 assays.