Learn More
Felix Storch Inc ACCUCOLD Performance series Pharma-Vac 4 cu.ft. ADA height purpose-built vaccine storage freezer -LHD
SDP
Catalog No. NC2403975
Encompass_Preferred
The AFZ5PVBIADA is a 4 cu.ft. all-freezer with a temperature range of -30C to -10C as recommended for a variety of vaccines and pharmaceuticals. It is designed for freestanding or recessed installation and sized just under 32' high.Accucold's Pharma-Vac Performance Series are upright freezers are designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC guidelines. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This model is equipped with remote alarm contacts in the rear and a 1/2' probe port on the right side to accept additional monitoring devices. A hospital grade cord with 'green dot' plug is also included for added safety in high traffic areas: https://www.accucold.com/product/AFZ5PVBIADA
Catalog No. NC2403975 Supplier Felix Storch Inc Supplier No. AFZ5PVBIADA
The AFZ5PVBIADA is a 4 cu.ft. all-freezer with a temperature range of -30C to -10C as recommended for a variety of vaccines and pharmaceuticals. It is designed for freestanding or recessed installation and sized just under 32' high.Accucold's Pharma-Vac Performance Series are upright freezers are designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC guidelines. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This model is equipped with remote alarm contacts in the rear and a 1/2' probe port on the right side to accept additional monitoring devices. A hospital grade cord with 'green dot' plug is also included for added safety in high traffic areas: https://www.accucold.com/product/AFZ5PVBIADA