Learn More
Spherotech ACCUCOUNT FLUORESCENT PART 5ML
Catalog No. NC0484660
Encompass_Preferred
AccuCount Fluorescent Particles, 10^6/mL, 7.0-7.9 um, 5 mL
Catalog No. NC0484660 Supplier Spherotech Supplier No. ACFP705
AccuCount Fluorescent Particles, 10^6/mL, 7.0-7.9 um, 5 mL